siRNA Details
| Gene Symbol: Tcrb-J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017898.1 | - | 4 | 513 | ||
| M11456.1 | - | 2 | 334 | |||
| M13676.1 | - | 2 | 307 | |||
| M16679.1 | - | 2 | 304 | |||
| M20877.1 | - | 1 | 256 | |||
| M20878.1 | - | 1 | 256 | |||
| U21402.1 | - | 58 | ||||
| U26785.1 | - | 64 | ||||
| U26791.1 | - | 1 | 49 | |||
| U26795.1 | - | 1 | 49 | |||
| U26796.1 | - | 1 | 49 | |||
| U46842.1 | - | 2 | 307 | |||
| X00438.1 | - | 2 | 329 | |||
| X02778.1 | - | 2 | 283 | |||
| X02779.1 | - | 2 | 307 | |||

Tcrb-J
Genomic Map