siRNA Details
| Gene Symbol: Tmem241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289666.1 | NP_001276595.1 | 11, 12 | 682 | ||
| NM_001289667.1 | NP_001276596.1 | 9, 10 | 522 | |||
| NM_178801.5 | NP_848916.2 | 11, 12 | 682 | |||
| GenBank mRNA | AK028973.1 | - | 232 | |||
| AK031517.1 | - | 12, 13 | 768 | |||
| AK038419.1 | - | 11, 12 | 660 | |||
| AK038472.1 | - | 11, 12 | 627 | |||
| AK144963.1 | - | 11, 12 | 684 | |||
| BC094382.1 | - | 11, 12 | 598 | 30605060 | ||
| BC137621.1 | - | 11, 12 | 612 | 8860640 | ||
| BC144788.1 | - | 9, 10 | 453 | |||

Tmem241
Genomic Map