siRNA Details
| Gene Symbol: Plekhm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183034.1 | NP_898855.1 | 4 | 939 | ||
| GenBank mRNA | AK154703.1 | - | 3 | 868 | ||
| AK155357.1 | - | 4 | 963 | |||
| AK157752.1 | - | 4 | 974 | |||
| BC053079.1 | - | 4 | 939 | 5702529 | ||

Plekhm1
Genomic Map