siRNA Details
Gene Symbol: Txlng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290776.1 | NP_001277705.1 | 11 | 1339 | ||
NM_178935.5 | NP_849266.1 | 10 | 1486 | |||
GenBank mRNA | AK030100.1 | - | 10 | 1433 | ||
AK031023.1 | - | 8 | 3025 | |||
AK031783.1 | - | 10 | 1400 | |||
AK044130.1 | - | 10 | 1435 | |||
BC117898.1 | - | 11 | 1280 | 40096960 | ||
BC117899.1 | - | 11 | 1280 | 40096967 | ||
DQ316984.1 | - | 10 | 1486 | |||
DQ320011.1 | - | 11 | 1339 |
