siRNA Details
| Gene Symbol: Txlng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290776.1 | NP_001277705.1 | 11 | 1437 | ||
| NM_178935.5 | NP_849266.1 | 10 | 1584 | |||
| GenBank mRNA | AK030100.1 | - | 10 | 1531 | ||
| AK031023.1 | - | 8 | 3123 | |||
| AK031783.1 | - | 10 | 1498 | |||
| AK044130.1 | - | 10 | 1533 | |||
| BC117898.1 | - | 11 | 1378 | 40096960 | ||
| BC117899.1 | - | 11 | 1378 | 40096967 | ||
| DQ320011.1 | - | 11 | 1437 | |||

Txlng
Genomic Map