siRNA Details
| Gene Symbol: Scarf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004157.2 | NP_001004157.2 | 3 | 274 | ||
| GenBank mRNA | AK133477.1 | - | 3 | 257 | ||
| AK220215.1 | - | 3 | 258 | |||
| BC057600.1 | - | 2 | 181 | |||
| BC080288.1 | - | 3 | 274 | 6411836 | ||

Scarf1
Genomic Map