siRNA Details
| Gene Symbol: Ccdc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159421.1 | NP_001152893.1 | 3, 4 | 681 | ||
| NM_001159422.1 | NP_001152894.1 | 4, 5 | 458 | |||
| NM_198645.2 | NP_941047.1 | 4, 5 | 807 | |||
| GenBank mRNA | AK135450.1 | - | 4, 5 | 387 | ||
| AV583692.1 | - | 4, 5 | 458 | |||
| BB686073.1 | - | 2, 3 | 122 | |||
| BC024406.1 | - | 4, 5 | 371 | |||
| BC024862.1 | - | 4, 5 | 782 | 5356370 | ||
| BC053690.1 | - | 4, 5 | 359 | |||
| CJ094333.1 | - | 3, 4 | 262 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001105875.

Ccdc58
Genomic Map