siRNA Details
Gene Symbol: Tmem151a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001885.1 | NP_001001885.1 | 2 | 1437 | ||
GenBank mRNA | AK139456.1 | - | 2 | 1491 | ||
AK144531.1 | - | 2 | 1453 | |||
AK155690.1 | - | 2 | 1456 | |||
AK163651.1 | - | 2 | 1454 | |||
BC072634.1 | - | 2 | 1437 | 6850125 |
