siRNA Details
| Gene Symbol: Dnah7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252070.1 | NP_001238999.1 | 7, 8 | 571 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnah7a| Gene Symbol: Dnah7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252070.1 | NP_001238999.1 | 7, 8 | 571 | ||