siRNA Details
| Gene Symbol: C330006A16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256521.1 | NP_001243450.1 | 2 | 812 | ||
| NM_001256522.1 | NP_001243451.1 | 2 | 652 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C330006A16Rik| Gene Symbol: C330006A16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256521.1 | NP_001243450.1 | 2 | 812 | ||
| NM_001256522.1 | NP_001243451.1 | 2 | 652 | |||