siRNA Details
Gene Symbol: Slc44a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081263.1 | NP_001074732.1 | 3 | 443 | ||
GenBank mRNA | BC086641.1 | - | 3 | 443 | 6417957 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc44a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081263.1 | NP_001074732.1 | 3 | 443 | ||
GenBank mRNA | BC086641.1 | - | 3 | 443 | 6417957 |