siRNA Details
| Gene Symbol: Gm1661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033774.2 | NP_001028946.1 | 3, 4 | 527 | ||
| NM_001145637.1 | NP_001139109.1 | 3, 4 | 527 | |||
| GenBank mRNA | AK132996.1 | - | 3, 4 | 528 | ||
| BC147060.1 | - | 1, 2 | 227 | 9056068 | ||
| BC147061.1 | - | 1, 2 | 227 | 9056035 | ||
| BC171980.1 | - | 1, 2 | 227 | |||

Gm1661
Genomic Map