siRNA Details
| Gene Symbol: Ythdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177680.3 | NP_808348.2 | 4 | 550 | ||
| GenBank mRNA | AK135309.1 | - | 4 | 806 | ||
| BC044650.1 | - | 3 | 422 | |||
| CA317046.1 | - | 3 | 362 | |||
| CN528111.1 | - | 4 | 472 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ythdc1| Gene Symbol: Ythdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177680.3 | NP_808348.2 | 4 | 550 | ||
| GenBank mRNA | AK135309.1 | - | 4 | 806 | ||
| BC044650.1 | - | 3 | 422 | |||
| CA317046.1 | - | 3 | 362 | |||
| CN528111.1 | - | 4 | 472 | |||