siRNA Details
| Gene Symbol: Fbxw15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199036.2 | NP_950201.2 | 9, 10 | 1321 | ||
| GenBank mRNA | AK087669.1 | - | 9, 10 | 1322 | ||
| BC099378.1 | - | 6, 7 | 760 | 30936553 | ||
| CO795020.1 | - | 3, 4 | 342 | |||
| DQ067445.1 | - | 9, 10 | 1289 | |||

Fbxw15
Genomic Map