siRNA Details
Gene Symbol: March11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177597.5 | NP_808265.2 | 4 | 1098 | ||
GenBank mRNA | AK162044.1 | - | 3 | 900 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: March11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177597.5 | NP_808265.2 | 4 | 1098 | ||
GenBank mRNA | AK162044.1 | - | 3 | 900 |