siRNA Details
| Gene Symbol: Serpinb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198680.2 | NP_941373.1 | 6 | 542 | ||
| GenBank mRNA | AK003220.1 | - | 6 | 543 | ||
| AY144685.1 | - | 5 | 502 | |||
| AY144686.1 | - | 5 | 502 | |||
| AY367774.1 | - | 5 | 486 | |||
| BC099584.1 | - | 6 | 539 | 30291007 | ||

Serpinb3b
Genomic Map