siRNA Details
| Gene Symbol: Gm10857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139587.1 | - | 1 | 1266 | ||
| AK139737.1 | - | 1 | 1280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10857| Gene Symbol: Gm10857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139587.1 | - | 1 | 1266 | ||
| AK139737.1 | - | 1 | 1280 | |||