siRNA Details
| Gene Symbol: Xirp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | KM273012.1 | - | 373 | |||
| Gene Symbol: Gm1322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033477.3 | NP_001028649.2 | 2 | 563 | ||
| GenBank mRNA | AK158464.1 | - | 2 | 564 | ||

Xirp2
Genomic Map