siRNA Details
| Gene Symbol: Gm1322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033477.3 | NP_001028649.2 | 2 | 615 | ||
| GenBank mRNA | AK158464.1 | - | 2 | 616 | ||
| Gene Symbol: Xirp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | KM273012.1 | - | 425 | |||

Gm1322
Genomic Map