siRNA Details
| Gene Symbol: Slc25a34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013780.1 | NP_001013802.1 | 1 | 271 | ||
| GenBank mRNA | AK085913.1 | - | 1 | 262 | ||
| AK142924.1 | - | 1 | 272 | |||
| BC089627.1 | - | 1 | 243 | 6758395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a34| Gene Symbol: Slc25a34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013780.1 | NP_001013802.1 | 1 | 271 | ||
| GenBank mRNA | AK085913.1 | - | 1 | 262 | ||
| AK142924.1 | - | 1 | 272 | |||
| BC089627.1 | - | 1 | 243 | 6758395 | ||