siRNA Details
Gene Symbol: Fam84b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162926.1 | NP_001156398.1 | 2 | 2412 | ||
GenBank mRNA | AK052388.1 | - | 1 | 289 | ||
AK139928.1 | - | 2 | 2293 | |||
AK162412.1 | - | 1 | 498 |
