siRNA Details
| Gene Symbol: Fam84b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162926.1 | NP_001156398.1 | 2 | 2213 | ||
| GenBank mRNA | AK052388.1 | - | 1 | 90 | ||
| AK139928.1 | - | 2 | 2094 | |||
| AK162412.1 | - | 1 | 299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam84b| Gene Symbol: Fam84b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162926.1 | NP_001156398.1 | 2 | 2213 | ||
| GenBank mRNA | AK052388.1 | - | 1 | 90 | ||
| AK139928.1 | - | 2 | 2094 | |||
| AK162412.1 | - | 1 | 299 | |||