siRNA Details
Gene Symbol: Gfral | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205844.3 | NP_995316.2 | 5 | 825 | ||
GenBank mRNA | AY457637.2 | - | 5 | 825 | ||
AY457638.2 | - | 5 | 825 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gfral | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205844.3 | NP_995316.2 | 5 | 825 | ||
GenBank mRNA | AY457637.2 | - | 5 | 825 | ||
AY457638.2 | - | 5 | 825 |