siRNA Details
| Gene Symbol: BC053393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025435.3 | NP_001020606.2 | 4 | 614 | ||
| GenBank mRNA | AK140690.1 | - | 4 | 469 | ||
| AK166706.1 | - | 4 | 491 | |||
| AK166838.1 | - | 4 | 491 | |||
| BC052651.1 | - | 4 | 605 | 30034019 | ||
| BC053393.1 | - | 2798 | ||||

BC053393
Genomic Map