siRNA Details
| Gene Symbol: Slc18b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183116.2 | NP_898939.2 | 2 | 244 | ||
| GenBank mRNA | AK183134.1 | - | 2 | 154 | ||
| CK625631.1 | - | 2 | 174 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc18b1| Gene Symbol: Slc18b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183116.2 | NP_898939.2 | 2 | 244 | ||
| GenBank mRNA | AK183134.1 | - | 2 | 154 | ||
| CK625631.1 | - | 2 | 174 | |||