siRNA Details
| Gene Symbol: Dnaic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034878.2 | NP_001030050.2 | 2 | 466 | ||
| GenBank mRNA | AK138639.1 | - | 2 | 393 | ||
| EF565270.1 | - | 1 | 82 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnaic2| Gene Symbol: Dnaic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034878.2 | NP_001030050.2 | 2 | 466 | ||
| GenBank mRNA | AK138639.1 | - | 2 | 393 | ||
| EF565270.1 | - | 1 | 82 | |||