siRNA Details
| Gene Symbol: Hhla1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145096.1 | NP_001138568.1 | 6 | 344 | ||
| GenBank mRNA | AK158329.1 | - | 6 | 346 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hhla1| Gene Symbol: Hhla1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145096.1 | NP_001138568.1 | 6 | 344 | ||
| GenBank mRNA | AK158329.1 | - | 6 | 346 | ||