siRNA Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081335.2 | NP_001074804.2 | 2 | 314 | ||
| GenBank mRNA | AK147675.1 | - | 2 | 762 | ||
| CJ110581.1 | - | 2 | 314 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cul9| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081335.2 | NP_001074804.2 | 2 | 314 | ||
| GenBank mRNA | AK147675.1 | - | 2 | 762 | ||
| CJ110581.1 | - | 2 | 314 | |||