siRNA Details
| Gene Symbol: 1700011I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029290.1 | NP_083566.1 | 7 | 665 | ||
| GenBank mRNA | AK005868.1 | - | 7 | 714 | ||
| BC048576.1 | - | 6 | 529 | |||
| CF981605.1 | - | 338 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700011I03Rik| Gene Symbol: 1700011I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029290.1 | NP_083566.1 | 7 | 665 | ||
| GenBank mRNA | AK005868.1 | - | 7 | 714 | ||
| BC048576.1 | - | 6 | 529 | |||
| CF981605.1 | - | 338 | ||||