siRNA Details
| Gene Symbol: Rorb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001043354.2 | NP_001036819.1 | 10 | 7134 | ||
| NM_001289921.1 | NP_001276850.1 | 11 | 6653 | |||
| NM_146095.4 | NP_666207.3 | 10 | 6600 | |||
| GenBank mRNA | AK159011.1 | - | 1 | 1403 | ||

Rorb
Genomic Map