siRNA Details
| Gene Symbol: Fam126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172513.3 | NP_766101.3 | 12 | 8397 | ||
| GenBank mRNA | AK048435.1 | - | 1 | 2415 | ||
| Gene Symbol: Gm15834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI875682.1 | - | 1 | 283 | ||
| AI891514.1 | - | 1 | 273 | |||
| AK048489.1 | - | 1 | 525 | |||
| AK053027.1 | - | 7 | 2483 | |||

Fam126b
Genomic Map