siRNA Details
Gene Symbol: Fam126b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172513.3 | NP_766101.3 | 12 | 8426 | ||
GenBank mRNA | AK048435.1 | - | 1 | 2444 |
Gene Symbol: Gm15834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI875682.1 | - | 1 | 254 | ||
AI891514.1 | - | 1 | 244 | |||
AK048489.1 | - | 1 | 496 | |||
AK053027.1 | - | 7 | 2454 |
