siRNA Details
| Gene Symbol: Creg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011804.2 | NP_035934.1 | 3 | 563 | ||
| GenBank mRNA | AF084524.1 | - | 3 | 563 | ||
| AK005388.1 | - | 3 | 584 | |||
| AK010947.1 | - | 3 | 580 | |||
| AK085844.1 | - | 3 | 375 | |||
| AK144101.1 | - | 3 | 582 | |||
| AK150006.1 | - | 4 | 689 | |||
| AK189906.1 | - | 3 | 274 | |||
| AK190952.1 | - | 3 | 272 | |||
| AK191906.1 | - | 3 | 272 | |||
| AK193253.1 | - | 3 | 278 | |||
| AK194678.1 | - | 3 | 273 | |||
| AK200846.1 | - | 3 | 272 | |||
| AK203453.1 | - | 3 | 275 | |||
| AK219109.1 | - | 3 | 284 | |||
| BC027426.1 | - | 3 | 563 | 3670649 | ||

Creg1
Genomic Map