siRNA Details
| Gene Symbol: Gm5544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033779.2 | NP_001028951.1 | 2 | 546 | ||
| GenBank mRNA | AK142329.1 | - | 2 | 547 | ||
| BC147112.1 | - | 1 | 380 | 9056126 | ||
| BC147113.1 | - | 1 | 380 | 9056128 | ||

Gm5544
Genomic Map