siRNA Details
Gene Symbol: 2610020H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004187.1 | NP_001004187.1 | 8 | 1123 | ||
NM_028129.2 | NP_082405.1 | 5 | 843 | |||
GenBank mRNA | AK011486.1 | - | 5 | 843 | ||
BC053445.1 | - | 8 | 1123 | 6336581 |
