siRNA Details
| Gene Symbol: 2610020H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004187.1 | NP_001004187.1 | 12 | 1541 | ||
| NM_028129.2 | NP_082405.1 | 11 | 1459 | |||
| GenBank mRNA | AK011486.1 | - | 11 | 1459 | ||
| BC053445.1 | - | 12 | 1541 | 6336581 | ||

2610020H08Rik
Genomic Map