siRNA Details
| Gene Symbol: Siva1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013929.2 | NP_038957.2 | 1, 2 | 249 | ||
| GenBank mRNA | AF033113.1 | - | 1, 2 | 110 | ||
| AF033114.1 | - | 1, 2 | 197 | |||
| AK010562.1 | - | 1, 2 | 175 | |||
| AK017570.1 | - | 1, 2 | 210 | |||
| AK153115.1 | - | 1, 2 | 159 | |||
| AK169200.1 | - | 1, 2 | 202 | |||
| AK184852.1 | - | 1, 2 | 54 | |||
| AK209128.1 | - | 1, 2 | 54 | |||
| BC012279.1 | - | 1, 2 | 231 | 4013026 | ||
| CB524656.1 | - | 1, 2 | 398 | |||

Siva1
Genomic Map