siRNA Details
| Gene Symbol: BC048562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004192.1 | NP_001004192.1 | 4 | 494 | ||
| GenBank mRNA | AK029636.1 | - | 4 | 495 | ||
| BC048562.1 | - | 4 | 494 | 6703719 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC048562| Gene Symbol: BC048562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004192.1 | NP_001004192.1 | 4 | 494 | ||
| GenBank mRNA | AK029636.1 | - | 4 | 495 | ||
| BC048562.1 | - | 4 | 494 | 6703719 | ||