siRNA Details
| Gene Symbol: Ly6g6f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163192.1 | NP_001156664.1 | 3 | 620 | ||
| GenBank mRNA | BC157908.1 | - | 3 | 620 | 9088110 | |
| BC172069.1 | - | 3 | 620 | 9054355 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly6g6f| Gene Symbol: Ly6g6f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163192.1 | NP_001156664.1 | 3 | 620 | ||
| GenBank mRNA | BC157908.1 | - | 3 | 620 | 9088110 | |
| BC172069.1 | - | 3 | 620 | 9054355 | ||