siRNA Details
| Gene Symbol: Gsdma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007461.1 | NP_001007462.1 | 6 | 800 | ||
| GenBank mRNA | AY679090.1 | - | 6 | 800 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsdma3| Gene Symbol: Gsdma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007461.1 | NP_001007462.1 | 6 | 800 | ||
| GenBank mRNA | AY679090.1 | - | 6 | 800 | ||