siRNA Details
| Gene Symbol: Itlnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007552.1 | NP_001007553.1 | 8 | 914 | ||
| GenBank mRNA | AY217760.1 | - | 8 | 914 | ||
| BC145376.1 | - | 8 | 867 | 9053934 | ||
| Gene Symbol: Itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC145377.1 | - | 8 | 870 | 9053935 | |
| BC150760.1 | - | 8 | 870 | 9087671 | ||

Itlnb
Genomic Map