siRNA Details
| Gene Symbol: Itlnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007552.1 | NP_001007553.1 | 8 | 954 | ||
| GenBank mRNA | AY217760.1 | - | 8 | 954 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itlnb| Gene Symbol: Itlnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007552.1 | NP_001007553.1 | 8 | 954 | ||
| GenBank mRNA | AY217760.1 | - | 8 | 954 | ||