siRNA Details
| Gene Symbol: a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015770.3 | NP_056585.2 | 2 | 211 | ||
| GenBank mRNA | BC009122.1 | - | 366 | |||
| L06451.1 | - | 2 | 250 | |||
| L06941.1 | - | 2 | 211 | |||
| S63413.1 | - | 358 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

a| Gene Symbol: a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015770.3 | NP_056585.2 | 2 | 211 | ||
| GenBank mRNA | BC009122.1 | - | 366 | |||
| L06451.1 | - | 2 | 250 | |||
| L06941.1 | - | 2 | 211 | |||
| S63413.1 | - | 358 | ||||