siRNA Details
| Gene Symbol: Sall2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244916.1 | NP_001231845.1 | 2 | 1414 | ||
| NM_015772.3 | NP_056587.2 | 2 | 1701 | |||
| GenBank mRNA | AB093233.1 | - | 2 | 1360 | ||
| AJ007396.1 | - | 2 | 1792 | |||
| AK077601.1 | - | 2 | 1328 | |||
| BC085361.1 | - | 2 | 1478 | 5706710 | ||

Sall2
Genomic Map