siRNA Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081069.1 | NP_001074538.1 | 10, 11 | 686 | ||
| GenBank mRNA | AF061934.1 | - | 9, 10 | 591 | ||
| AK039314.1 | - | 9, 10 | 1308 | |||
| AK080707.1 | - | 10, 11 | 777 | |||
| AK161551.1 | - | 6, 7 | 745 | |||
| BC049910.1 | - | 8, 9 | 1390 | |||

Rgs11
Genomic Map