siRNA Details
| Gene Symbol: Tmod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038710.1 | NP_001033799.1 | 8 | 1190 | ||
| NM_016711.3 | NP_057920.2 | 8 | 1126 | |||
| GenBank mRNA | AF177170.1 | - | 8 | 1075 | ||
| AF237629.1 | - | 9 | 1034 | |||
| AK017072.1 | - | 8 | 1174 | |||
| AK076562.1 | - | 8 | 1140 | |||
| AK079077.1 | - | 8 | 970 | |||
| AK147581.1 | - | 8 | 1122 | |||
| AK164178.1 | - | 8 | 972 | |||
| BC052227.1 | - | 8 | 1190 | |||
| BC061124.1 | - | 8 | 956 | 6531518 | ||

Tmod2
Genomic Map