siRNA Details
| Gene Symbol: Preb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294302.1 | NP_001281231.1 | 7 | 1196 | ||
| NM_016703.3 | NP_057912.2 | 7 | 1196 | |||
| GenBank mRNA | AF150808.1 | - | 7 | 1099 | ||
| AF193017.1 | - | 7 | 1108 | |||
| AK013330.1 | - | 7 | 1150 | |||
| AK029838.1 | - | 7 | 1132 | |||
| AK036866.1 | - | 7 | 1134 | |||
| AK151288.1 | - | 7 | 1178 | |||
| AK179426.1 | - | 2 | 152 | |||
| AK202976.1 | - | 2 | 91 | |||
| AK206719.1 | - | 2 | 152 | |||
| BC017527.1 | - | 7 | 1099 | 4921119 | ||

Preb
Genomic Map