siRNA Details
| Gene Symbol: Reep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178608.4 | NP_848723.1 | 3, 4 | 473 | ||
| GenBank mRNA | AK035238.1 | - | 3, 4 | 307 | ||
| AK045056.1 | - | 3, 4 | 295 | |||
| AK053158.1 | - | 3, 4 | 291 | |||
| AK136916.1 | - | 3, 4 | 252 | |||
| AY562229.1 | - | 3, 4 | 184 | |||
| BC046826.1 | - | 3, 4 | 244 | 6417744 | ||
| CX239705.1 | - | 3, 4 | 473 | |||

Reep1
Genomic Map