siRNA Details
Gene Symbol: Adgra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177469.3 | NP_803420.2 | 8 | 2216 | ||
GenBank mRNA | AB096262.1 | - | 6 | 1279 | ||
AK034188.1 | - | 7 | 1779 | |||
AK082207.1 | - | 8 | 2095 | |||
BC056493.1 | - | 6 | 1500 | 5704932 | ||
BC094311.1 | - | 3 | 764 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
