siRNA Details
| Gene Symbol: Adgra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177469.3 | NP_803420.2 | 5, 6 | 1399 | ||
| GenBank mRNA | AB096262.1 | - | 3, 4 | 462 | ||
| AK034188.1 | - | 4, 5 | 962 | |||
| AK082207.1 | - | 5, 6 | 1278 | |||
| BC056493.1 | - | 3, 4 | 683 | 5704932 | ||
More Information
Set Membership: |
|

Adgra1
Genomic Map